Ethylenediamine-N,N'-diacetic-N,N'-dipropionic acid hydrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: EDDADP

E0288-1G
TCIAE0288-1GEA 1500 NOK
TCIAE0288-1G
Ethylenediamine-N,N'-diacetic-N,N'-dipropionic acid hydrate ≥98.0% (by titrimetric analysis)
Ethylenediamine-N,N'-diacetic-N,N'-dipropionic acid hydrate
Formel: C₁₂H₂₀N₂O₈
MDL nummer: MFCD00002768
CAS-nummer: 32701-19-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in hot Water almost transparency
Water max. 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR