N-Etylacetamid ≥99.0% (by GC)

Leverandør: TCI

E0324-25ML
TCIAE0324-25MLEA 795 NOK
TCIAE0324-25ML
N-Etylacetamid ≥99.0% (by GC)
N-Etylacetamid
Formel: C₄H₉NO
MW: 87,12 g/mol
Kokepunkt: 206°C
Smeltepunkt: –32°C
Flash punkt: 110°C
MDL nummer: MFCD00009029
CAS-nummer: 625-50-3
EINECS: 210-896-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45e846f11d49feb3e5632cc01743b11ee0deeff4&vid=45e846f11d49feb3e5632cc01743b11ee0deeff4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR