9,10-Epoxy-1-decene ≥96.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1,2-Epoxy-9-decene

E0411-5ML
TCIAE0411-5MLEA 1325 NOK
TCIAE0411-5ML
9,10-Epoxy-1-decene ≥96.0% (by GC)
9,10-Epoxy-1-decene
Formel: C₁₀H₁₈O
MW: 154,25 g/mol
Kokepunkt: 80°C
Flash punkt: 78°C
MDL nummer: MFCD00069427
CAS-nummer: 85721-25-1
EINECS: 288-437-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a2f970119383d4ae7996b7bbeaea5bc729338b&vid=39a2f970119383d4ae7996b7bbeaea5bc729338b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR