all cis-5,8,11,14,17-Eicosapentaenoic acid ≥95.0% (by GC)

Leverandør: TCI

E0441-100MG E0441-500MG
TCIAE0441-100MGEA 2225 NOK
TCIAE0441-100MG TCIAE0441-500MG
all cis-5,8,11,14,17-Eicosapentaenoic acid ≥95.0% (by GC)
all cis-5,8,11,14,17-Eicosapentaenoic acid
Formel: C₂₀H₃₀O₂
Flash punkt: 93°C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00065716
CAS-nummer: 10417-94-4
Merck Indeks: 14,03531

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR