cis-Epoxysuccinic acid ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: cis-2,3-Oxiranedicarboxylic Acid

E0449-25G E0449-5G
TCIAE0449-25GEA 3050 NOK
TCIAE0449-25G TCIAE0449-5G
cis-Epoxysuccinic acid ≥97.0% (by GC, titration analysis)
cis-Epoxysuccinic acid
Formel: C₄H₄O₅
MDL nummer: MFCD00191645
CAS-nummer: 16533-72-5
EINECS: 240-604-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 149.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR