Ethyl-2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 2-(2-Carbethoxyphenyl)-5,5-dimethyl-1,3,2-dioxaborinane, 2-(2-Ethoxycarbonylphenyl)-5,5-dimethyl-1,3,2-dioxaborinane, 2-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzoic Acid Ethyl Ester, HSL-IN-3, Ethyl 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate

E0667-1G
TCIAE0667-1GEA 2350 NOK
TCIAE0667-1G
Ethyl-2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate ≥98.0% (by titrimetric analysis)
Ethyl-2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate
Formel: C₁₄H₁₉BO₄
MW: 262,11 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD08276372
CAS-nummer: 346656-34-6

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR