trans-4-(4-Ethylcyclohexyl)benzonitrile ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 4-(trans-4-Ethylcyclohexyl)benzonitrile

E0703-5G
TCIAE0703-5GEA 540 NOK
TCIAE0703-5G
trans-4-(4-Ethylcyclohexyl)benzonitrile ≥98.0% (by GC)
trans-4-(4-Ethylcyclohexyl)benzonitrile
Formel: C₁₅H₁₉N
MW: 213,32 g/mol
MDL nummer: MFCD09750932
CAS-nummer: 72928-54-2
EINECS: 277-084-2
FN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce201663016f610d4b10f3364f8e0a03695ea56f&vid=ce201663016f610d4b10f3364f8e0a03695ea56f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR