7-Ethyl-10-hydroxycamptothecin ≥98.0% (by HPLC)

Leverandør: TCI
Danger

E0748-100MG E0748-1G
TCIAE0748-100MGEA 1175 NOK
TCIAE0748-100MG TCIAE0748-1G
7-Ethyl-10-hydroxycamptothecin ≥98.0% (by HPLC)
7-Ethyl-10-hydroxycamptothecin
Formel: C₂₂H₂₀N₂O₅
Smeltepunkt: 217°C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD06762720
CAS-nummer: 86639-52-3
FN: 1544
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60bbfada20e8f392358c9ee4542b911fa8a4b098&vid=1edad1245472dcce117eeea0ddf5098cb8291045&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 94.0 %
Specific rotation [a]20/D +20.0 to +30.0 deg (C=0.2, THF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR