Ethylmagnesium chloride ∼1.0 mol/L in THF activated with Zinc Chloride (∼10mol%), activated with zinc chloride (ca. 10mol%)

Leverandør: TCI
Danger

E0778-250G
TCIAE0778-250GEA 1325 NOK
TCIAE0778-250G
Ethylmagnesium chloride ∼1.0 mol/L in THF activated with Zinc Chloride (∼10mol%), activated with zinc chloride (ca. 10mol%)
Ethylmagnesium chloride
Formel: C₂H₅ClMg
MDL nummer: MFCD00000472
CAS-nummer: 2386-64-3
EINECS: 219-206-9
FN: 3399
ADR: 4.3,II

Bestill nå

Spesifikasjon Testresultater

Concentration (Neutralization titration) 9.0 to 11.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR