Ethanolamine thioglycolate ∼85% in water

Leverandør: TCI
Warning

Synonymer: Ethanolamine Mercaptoacetate, Monoethanolamine Thioglycolate

E0813-25G E0813-500G
TCIAE0813-25GEA 220 NOK
TCIAE0813-25G TCIAE0813-500G
Ethanolamine thioglycolate ∼85% in water
Ethanolamine thioglycolate
Formel: C₄H₁₁NO₃S
MW: 153.20 g/mol
MDL nummer: MFCD00050789
CAS-nummer: 126-97-6
EINECS: 204-815-4

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Concentration (Iodometric titration) 83.0 to 88.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR