β-Estradiol-17-valerate ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: beta-Estradiol 17-Valerate

E0876-1G E0876-5G
TCIAE0876-1GEA 1150 NOK
TCIAE0876-1G TCIAE0876-5G
β-Estradiol-17-valerate ≥98.0% (by GC)
β-Estradiol-17-valerate
Formel: C₂₃H₃₂O₃
MW: 356.506 g/mol
MDL nummer: MFCD00056541
CAS-nummer: 979-32-8
EINECS: 213-559-2
Merck Indeks: 14,03703

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 146.0 to 150.0 deg-C
Specific rotation [a]20/D +41.0 to +44.0 deg (C=2, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR