4-Ethylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-Ethylcyclohexanecarboxylic acid

E0966-5G
TCIAE0966-5GEA 2975 NOK
TCIAE0966-5G
4-Ethylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
4-Ethylcyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₉H₁₆O₂
MDL nummer: MFCD01568857
CAS-nummer: 91328-77-7

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR