beta-Estradiol 17-Heptanoate 98,0 GC_ASSAY_METHOD

Leverandør: TCI
Warning

Synonymer: beta-Estradiol 17-Enanthate

E1151-1G
TCIAE1151-1GEA 905 NOK
TCIAE1151-1G
beta-Estradiol 17-Heptanoate 98,0 GC_ASSAY_METHOD
beta-Estradiol 17-Heptanoate
Formel: C₂₅H₃₆O₃
MW: 384.56 g/mol
MDL nummer: MFCD00056542
CAS-nummer: 4956-37-0
EINECS: 225-599-8
Merck Indeks: 14,03703

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C
Specific rotation [a]20/D +42.0 to +46.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR