Flavianic acid dihydrate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 2,4-Dinitro-1-naphthol-7-sulphonic acid, 8-Hydroxy-5,7-dinitronaphthalene-2-sulphonic acid, 8-Hydroxy-5,7-dinitro-2-naphthalenesulphonic acid

F0012-25G
TCIAF0012-25GEA 610 NOK
TCIAF0012-25G
Flavianic acid dihydrate ≥98.0% (by HPLC, titration analysis)
Flavianic acid dihydrate
Formel: C₁₀H₆N₂O₈S
MW: 314,22 g/mol
MDL nummer: MFCD00046407
CAS-nummer: 483-84-1
EINECS: 207-600-3
FN: 3261
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Melting point 144.0 to 154.0 deg-C
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR