Acid red 88

Leverandør: TCI

Synonymer: Echt Rot, Fast Red A, Roccelline

F0087-25G
TCIAF0087-25GEA 1100 NOK
TCIAF0087-25G
Acid red 88
Acid red 88
Formel: C₂₀H₁₄N₂O₄S
MDL nummer: MFCD00004070
CAS-nummer: 1658-56-6
EINECS: 216-760-3

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Lambda max. 503.0 to 507.0 nm (H2O)
Absorbance (E1%1cm) min. 400 (H2O, 503.0 to 507.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR