N-Formyl-DL-alanine ≥98.0% (by titrimetric analysis)

Leverandør: TCI

F0107-1G
TCIAF0107-1GEA 1225 NOK
TCIAF0107-1G
N-Formyl-DL-alanine ≥98.0% (by titrimetric analysis)
N-Formyl-DL-alanine
Formel: C₄H₇NO₃
MDL nummer: MFCD00063285
CAS-nummer: 5893-10-7
EINECS: 227-574-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 145.0 to 150.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR