Food Red No. 104

Leverandør: TCI

F0142-25G
TCIAF0142-25GEA 1050 NOK
TCIAF0142-25G
Food Red No. 104
Standarder Mat standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ff0e3128b96689ee7a014b93fa2cb11f84f6383&vid=b728c5c9498f97868dd1f8018f24172b8578c75e&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse Food Red No. 104
Environmentally Preferable
CAS-nr 18472-87-2
Formel C₂₀H₂Br₄Cl₄Na₂O₅

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR