Food Yellow No. 4

Leverandør: TCI

F0144-25G
TCIAF0144-25GEA 825 NOK
TCIAF0144-25G
Food Yellow No. 4
Standarder Mat standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45cded3a44963baed1564aba1f89f32b8e930150&vid=be6b3589242abe0551f29a58b1f702cc569e7a48&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse Food Yellow No. 4
Environmentally Preferable
CAS-nr 1934-21-0
Formel C₁₆H₉N₄Na₃O₉S₂

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR