Food Blue No. 2

Leverandør: TCI

F0148-25G
TCIAF0148-25GEA 945 NOK
TCIAF0148-25G
Food Blue No. 2
Standarder Mat standarder

Food Blue No. 1 consists mainly of disodium 2-[bis[4- [N-ethyl-N-(3-sulfonatophenylmethyl)amino]phenyl]methylio]benzenesulfonate, (C37H34N2Na2O9S3).

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35bf6b532e42deb1d9201956fab8763e1279c96b&vid=35bf6b532e42deb1d9201956fab8763e1279c96b&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse Food Blue No. 2
Environmentally Preferable
CAS-nr 860-22-0
Formel C₁₆H₈N₂Na₂O₈S₂

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR