D(+)-Fenkon ≥98.0% (by GC)

Leverandør: TCI

F0163-25ML
TCIAF0163-25MLEA 395 NOK
TCIAF0163-25ML
D(+)-Fenkon ≥98.0% (by GC)
D(+)-Fenkon
Formel: C₁₀H₁₆O
Kokepunkt: 203 °C (1013 hPa)
Tetthet: 0,9449 g/cm³ (20 °C)
Flash punkt: 63°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070689
CAS-nummer: 4695-62-9
EINECS: 225-160-0
ADR: 3,III
Merck Indeks: 14,03968
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab1412f91b2accc84ca60a5f16342ce77af088c9&vid=ab1412f91b2accc84ca60a5f16342ce77af088c9&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 90.0 ee%
Specific rotation [a]20/D +55.0 to +63.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR