2-Fluoro-DL-phenylalanine ≥97.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 2-Amino-3-(2-fluorophenyl)propanoic acid

F0170-1G
TCIAF0170-1GEA 460 NOK
TCIAF0170-1G
2-Fluoro-DL-phenylalanine ≥97.0% (by titrimetric analysis)
2-Fluoro-DL-phenylalanine
Formel: C₉H₁₀FNO₂
MW: 183,18 g/mol
MDL nummer: MFCD00004271
CAS-nummer: 2629-55-2
EINECS: 220-105-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3ac243fe8660987839ac518e47fd2152d5e815e&vid=f3ac243fe8660987839ac518e47fd2152d5e815e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 236.0 to 242.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR