5-Fluoro-o-toluidine ≥98.0% (by GC)

Leverandør: TCI
Warning

F0232-25G F0232-5G
TCIAF0232-25GEA 1025 NOK
TCIAF0232-25G TCIAF0232-5G
5-Fluoro-o-toluidine ≥98.0% (by GC)
5-Fluoro-o-toluidine
Formel: C₇H₈FN
Kokepunkt: 83°C
Smeltepunkt: 36…38 °C
Tetthet: 1,13 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007764
CAS-nummer: 367-29-3
EINECS: 206-689-6
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89c09f2885b8e56cca6dd71455516d1b7df5e13c&vid=89c09f2885b8e56cca6dd71455516d1b7df5e13c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR