Fluorescein diacetate ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: Di-O-acetylfluorescein, 3,6-Diacetoxyfluoran, 3-Oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl diacetate

F0240-5G
TCIAF0240-5GEA 785 NOK
TCIAF0240-5G
Fluorescein diacetate ≥98.0% (by HPLC)
Fluorescein diacetate
Formel: C₂₄H₁₆O₇
MDL nummer: MFCD00005062
CAS-nummer: 596-09-8
EINECS: 209-877-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0499321d040564b9ddb5846decada52f9c283f87&vid=0499321d040564b9ddb5846decada52f9c283f87&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystaline
Purity (HPLC) min. 98.0 area%
Melting point 204.0 to 210.0 deg-C
Solubility in Chloroform almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR