Fenoprofen calcium salt dihydrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

F0372-25G
TCIAF0372-25GEA 1850 NOK
TCIAF0372-25G
Fenoprofen calcium salt dihydrate ≥98.0% (by titrimetric analysis)
Fenoprofen calcium salt dihydrate
Formel: C₃₀H₃₀CaO₈
MDL nummer: MFCD00242722
CAS-nummer: 71720-56-4
Merck Indeks: 14,03980

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in EtOH almost transparency
Water 6.0 to 6.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR