2-(Furfurylthio)ethylamine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

F0378-25G F0378-5G
TCIAF0378-25GEA 2225 NOK
TCIAF0378-25G TCIAF0378-5G
2-(Furfurylthio)ethylamine ≥98.0% (by GC, titration analysis)
2-(Furfurylthio)ethylamine
Formel: C₇H₁₁NOS
Kokepunkt: 100°C
MDL nummer: MFCD01075029
CAS-nummer: 36415-21-1
EINECS: 253-024-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adaf2777ccc86a11091791addd9f12714fcba9aa&vid=adaf2777ccc86a11091791addd9f12714fcba9aa&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR