5-Fluorocytidine ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-fluoropyrimidin-2(1H)-one

F0534-1G
TCIAF0534-1GEA 715 NOK
TCIAF0534-1G
5-Fluorocytidine ≥97.0% (by titrimetric analysis)
5-Fluorocytidine
Formel: C₉H₁₂FN₃O₅
MW: 261,21 g/mol
Smeltepunkt: 193°C
MDL nummer: MFCD00210953
CAS-nummer: 2341-22-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D +54.0 to +58.0 deg (C=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR