Fluorescein 6-isothiocyanate (isomer II) ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 3',6'-Dihydroxy-6-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 6-FITC, Fluorescein Isothiocyanate, 6-Isothiocyanatofluorescein, Fluorescein-6-isothiocyanate

F0783-100MG
TCIAF0783-100MGEA 2350 NOK
TCIAF0783-100MG
Fluorescein 6-isothiocyanate (isomer II) ≥97.0% (by HPLC, titration analysis)
Fluorescein 6-isothiocyanate (isomer II)
Formel: C₂₁H₁₁NO₅S
MW: 389.38 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00041838
CAS-nummer: 18861-78-4

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to Amber powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR