Fluorescein isothiocyanate dextran, M.W. ∼ 10000

Leverandør: TCI

Synonymer: FITC Dextran

F0918-100MG
TCIAF0918-100MGEA 1400 NOK
TCIAF0918-100MG
Fluorescein isothiocyanate dextran, M.W. ∼ 10000
Fluorescein isothiocyanate dextran
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00131092
CAS-nummer: 60842-46-8

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange powder to crystaline
Drying loss max. 7.0 %
Functionality test to pass test

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR