N-Formyl-L-Methionyl-L-Leucyl-L-Phenylalanine

Leverandør: TCI

F1158-25MG F1158-100MG
TCIAF1158-25MGEA 715 NOK
TCIAF1158-25MG TCIAF1158-100MG
N-Formyl-L-Methionyl-L-Leucyl-L-Phenylalanine
Proteiner

  • Specific rotation [a]20/D: –8,0 to –10,0° (C=1, acetic acid)

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ebbd0fb785104c11c641766674f51dd65ad9f47&vid=9911053de2024b99363da3f3e47f097b35dbee26&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Protein/peptide name N-Formyl-L-methionyl-L-leucyl-L-phenylalanine
Environmentally Preferable
Protein synonyms N-Formyl-Met-Leu-Phe|fMLP|fMLF
Arter
Kilde
CAS-nr 59880-97-6
MDL No. MFCD00036780
Renhet 98,0% (HPLC)
Skjema White to almost white powder to crystal
Molekylvekt 437,56 g/mol
Molekylformel C₂₁H₃₁N₃O₅S
Lagringsforhold Freezer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR