Fidaxomicin ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: Lipiarmycin A3, Tiacumicin B, (2R,3S,4S,5S,6R)-6-(((3E,5E,8S,9E,11S,12R,13E,15E,18S)-12-(((2R,3S,4R,5S)-3,4-Dihydroxy-5-(isobutyryloxy)-6,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-11-ethyl-8-hydroxy-18-((R)-1-hydroxyethyl)-9,13,15-trimethyl-2-oxooxacyclooctadeca-3,5,9,13,15-pentaen-3-yl)methoxy)-4-hydroxy-5-methoxy-2-methyltetrahydro-2H-pyran-3-yl 3,5-dichloro-2-ethyl-4,6-dihydroxybenzoate, Clostomicin B1

F1216-100MG F1216-25MG
TCIAF1216-100MGEA 4625 NOK
TCIAF1216-100MG TCIAF1216-25MG
Fidaxomicin ≥98.0% (by HPLC)
Fidaxomicin
Formel: C₅₂H₇₄Cl₂O₁₈
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD27976367
CAS-nummer: 873857-62-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR