5-FAM-Alkyne ≥97.0% (by HPLC)

Leverandør: TCI

Synonymer: 3',6'-Dihydroxy-3-oxo-N-(prop-2-yn-1-yl)-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxamide

F1222-25MG
TCIAF1222-25MGEA 5300 NOK
TCIAF1222-25MG
5-FAM-Alkyne ≥97.0% (by HPLC)
5-FAM-Alkyne
Formel: C₂₄H₁₅NO₆
Lagringstemperatur: Fryseskap
MDL nummer: MFCD28505571
CAS-nummer: 510758-19-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR