Monoolein 40,00% (by GC)

Leverandør: TCI

Synonymer: 9-Octadecenoicacid(Z)-,2,3-dihydroxypropylester, 1-(cis-9-Octadecenoyl)-rac-glycerol, Glyceryl cis-9-octadecenate hygroscopic, DL-α-Monoolein, rac-Glycerol 1-monooleate, 1-Oleoyl-rac-glycerol, 1-Mono(cis-9-octadecenoyl)-rac-glycerol, 1-Monleoyl-rac-glycerol, Glyceryl monooleate

G0082-25G G0082-500G
TCIAG0082-25GEA 230 NOK
TCIAG0082-25G TCIAG0082-500G
Monoolein 40,00% (by GC)
Monoolein
Formel: C₂₁H₄₀O₄
Flash punkt: 180°C
MDL nummer: MFCD00042735
CAS-nummer: 111-03-5
EINECS: 203-827-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange clear liquid
Purity (GC) min. 40.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR