Monopalmitin 95,0 GC_ASSAY_METHOD

Leverandør: TCI

Synonymer: Glycerol 1-palmitate, Monopalmitin(Chunks or pellets), Glycerol alpha-Monopalmitate, 1-Palmitoylglycerol, 1-Monopalmitoyl-rac-Glycerol

G0083-25G
TCIAG0083-25GEA 1175 NOK
TCIAG0083-25G
Monopalmitin 95,0 GC_ASSAY_METHOD
Monopalmitin
Formel: C₁₉H₃₈O₄
MW: 330.51 g/mol
MDL nummer: MFCD00042734
CAS-nummer: 542-44-9
EINECS: 208-812-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 73.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR