Monostearin >60.0% (by GC)

Leverandør: TCI

Synonymer: 2,3-Dihydroxypropyl stearate, 1-Stearoyl-rac-glycerol

G0085-25G G0085-500G
TCIAG0085-25GEA 220 NOK
TCIAG0085-25G TCIAG0085-500G
Monostearin >60.0% (by GC)
Monostearin
Formel: C₂₁H₄₂O₄
MDL nummer: MFCD00036186
CAS-nummer: 123-94-4
EINECS: 204-664-4
Merck Indeks: 14,04489

Bestill nå

Spesifikasjon Testresultater

Appearance White powder to crystal
Purity (GC) min. 60.0 % (total of isomer)
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR