Glyoxal 39% ∼8.8 mol/L in water

Leverandør: TCI
Danger

Synonymer: Ethandial

G0152-25G G0152-500G
TCIAG0152-25GEA 220 NOK
TCIAG0152-25G TCIAG0152-500G
Glyoxal 39% ∼8.8 mol/L in water
Glyoxal
Formel: C₂H₂O₂
MW: 58.04 g/mol
Kokepunkt: 51
Tetthet: 1.14
MDL nummer: MFCD00006957
CAS-nummer: 107-22-2
EINECS: 203-474-9
Merck Indeks: 14,04509

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Concentration (Neutralization titration) 39.0 to 42.0 w/w%
Acid Value max. 10.0

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR