Galβ(1-3)GalNAc-β-pNP ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: 4-Nitrophenyl 2-Acetamido-2-deoxy-3-O-(β-D-galactopyranosyl)-β-D-galactopyranoside

G0344-5MG
TCIAG0344-5MGEA 2925 NOK
TCIAG0344-5MG
Galβ(1-3)GalNAc-β-pNP ≥98.0% (by HPLC)
Gal beta(1-3)GalNAc-beta-pNP
Formel: C₂₀H₂₈N₂O₁₃
MW: 504,45 g/mol
Lagringstemperatur: Fryseskap
CAS-nummer: 59837-15-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR