Glyoxylic acid ∼50% ∼9 mol/L in water

Leverandør: TCI
Danger

G0366-25ML G0366-500ML
TCIAG0366-25MLEA 220 NOK
TCIAG0366-25ML TCIAG0366-500ML
Glyoxylic acid ∼50% ∼9 mol/L in water
Glyoxylic acid
Formel: C₂H₂O₃
MDL nummer: MFCD00006958
CAS-nummer: 298-12-4
EINECS: 206-058-5
FN: 3265
ADR: 8,III
Merck Indeks: 14,04511
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2fb5238013e2c9070938782ca61c4713047711a&vid=bcdfec2e514e4d8308e67f8096a4e00111536140&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Concentration (Neutralization titration) 50.0 to 55.0 w/w% (calcd. on exclusive of Oxalic acid)
Oxalic Acid max. 4.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR