1-Heneicosanol ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Heneicosanol, Heneicosyl alcohol, 1-Heneicosanol

H0011-10G
TCIAH0011-10GEA 12800 NOK
TCIAH0011-10G
1-Heneicosanol ≥98.0% (by GC)
1-Heneicosanol
Formel: H₃C(CH₂)₁₉CH₂OH
Kokepunkt: 170 °C (1013 hPa)
Smeltepunkt: 68…69 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00062834
CAS-nummer: 15594-90-8
EINECS: 239-673-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=385bda475a4013db31aac5680bf3f8a7b0b79bc6&vid=385bda475a4013db31aac5680bf3f8a7b0b79bc6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 70.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR