(±)-3-Heptanol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-Heptyl alcohol, 3-Heptanol, Heptan-3-ol, Butylethylcarbinol, (±)-Ethyl butyl carbinol, (±)-Butyl ethyl carbinol, (±)-3-Hydroxy heptane

H0035-25ML H0035-500ML
TCIAH0035-25MLEA 660 NOK
TCIAH0035-25ML TCIAH0035-500ML
(±)-3-Heptanol ≥98.0% (by GC)
(±)-3-Heptanol
Formel: C₇H₁₆O
Kokepunkt: 156°C
Smeltepunkt: –70 °C
Tetthet: 0,837 g/cm³ (20 °C)
Flash punkt: 60°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004586
CAS-nummer: 589-82-2
EINECS: 209-661-1
FN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=748a61bdc580a5cb329259f9747bc9a151618835&vid=f74db19b9c6da94b8e88755f773e99ec750be3b1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR