Homatropine hydrobromide ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: DL-Homatropine Hydrobromide

H0155-1G
TCIAH0155-1GEA 595 NOK
TCIAH0155-1G
Homatropine hydrobromide ≥99.0% (by titrimetric analysis)
Homatropine hydrobromide
Formel: C₁₆H₂₁NO₃
MDL nummer: MFCD00012676
CAS-nummer: 51-56-9
EINECS: 200-105-3
FN: 1544
ADR: 6.1,III
Merck Indeks: 14,04730

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR