4-(Hydroxyamino)quinoline-N-oxide ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: N-(4-Quinolyl)hydroxylamine 1'-Oxide

H0515-1G H0515-5G
TCIAH0515-1GEA 1400 NOK
TCIAH0515-1G TCIAH0515-5G
4-(Hydroxyamino)quinoline-N-oxide ≥97.0% (by titrimetric analysis)
4-(Hydroxyamino)quinoline-N-oxide
Formel: C₉H₈N₂O₂
Smeltepunkt: 214°C
MDL nummer: MFCD00059752
CAS-nummer: 4637-56-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Amber powder to lump
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR