Lithium-DL-2-hydroxybutyrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: DL-2-Hydroxybutyric Acid Lithium Salt

H0580-25G
TCIAH0580-25GEA 1700 NOK
TCIAH0580-25G
Lithium-DL-2-hydroxybutyrate ≥98.0% (by titrimetric analysis)
Lithium-DL-2-hydroxybutyrate
Formel: C₄H₈O₃
MW: 110.04 g/mol
MDL nummer: MFCD00040991
CAS-nummer: 381716-41-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR