cis-3-Hexenyl-cis-3-hexenoate ≥95.0% (by GC)

Leverandør: TCI

Synonymer: cis-3-Hexenoic Acid cis-3-Hexenyl Ester

H0677-5ML
TCIAH0677-5MLEA 915 NOK
TCIAH0677-5ML
cis-3-Hexenyl-cis-3-hexenoate ≥95.0% (by GC)
cis-3-Hexenyl-cis-3-hexenoate
Formel: C₁₂H₂₀O₂
MW: 196,29 g/mol
Flash punkt: 104°C
MDL nummer: MFCD00036652
CAS-nummer: 61444-38-0
EINECS: 262-797-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f64369e7b6adef9c32b6c68e5e6e0dcb4eaee3ff&vid=f64369e7b6adef9c32b6c68e5e6e0dcb4eaee3ff&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR