4-Hydroxypiperidine hydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 4-Piperidinol Hydrochloride, Piperidin-4-ol hydrochloride

H0686-25G
TCIAH0686-25GEA 1500 NOK
TCIAH0686-25G
4-Hydroxypiperidine hydrochloride ≥98.0% (by titrimetric analysis)
4-Hydroxypiperidine hydrochloride
Formel: C₅H₁₁NO
MW: 137,61 g/mol
MDL nummer: MFCD00035298
CAS-nummer: 5382-17-2
EINECS: 226-374-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystaline
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 154.0 to 157.0 deg-C
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR