4-(4-Heptylphenyl)benzoic acid ≥95.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 4-Heptylbiphenyl-4'-carboxylic Acid

H0709-5G
TCIAH0709-5GEA 1175 NOK
TCIAH0709-5G
4-(4-Heptylphenyl)benzoic acid ≥95.0% (by GC)
4-(4-Heptylphenyl)benzoic acid
Formel: C₂₀H₂₄O₂
MW: 296,41 g/mol
MDL nummer: MFCD00143231
CAS-nummer: 58573-94-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Solubility in hot EtOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR