α,α-Diphenyl-N-methyl-L-prolinol ≥98.0% (by GC)

Leverandør: TCI
Warning

H0768-1G H0768-100MG H0768-5G
TCIAH0768-1GEA 2325 NOK
TCIAH0768-1G TCIAH0768-100MG TCIAH0768-5G
α,α-Diphenyl-N-methyl-L-prolinol ≥98.0% (by GC)
α,α-Diphenyl-N-methyl-L-prolinol
Formel: C₁₈H₂₁NO
MDL nummer: MFCD00145245
CAS-nummer: 110529-22-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=896da237e6a0bc078273f140734daccaedea32e4&vid=896da237e6a0bc078273f140734daccaedea32e4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystaline
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C
Specific rotation [a]20/D +52.0 to +57.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR