(S)-(+)-3-Hydroxy-2,2-dimethylcyclohexanone ≥95.0% (by GC)

Leverandør: TCI

H0887-100MG
TCIAH0887-100MGEA 1575 NOK
TCIAH0887-100MG
(S)-(+)-3-Hydroxy-2,2-dimethylcyclohexanone ≥95.0% (by GC)
(S)-(+)-3-Hydroxy-2,2-dimethylcyclohexanone
Formel: C₈H₁₄O₂
MW: 142.20 g/mol
MDL nummer: MFCD00671542
CAS-nummer: 87655-21-8

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 95.0 %
Specific rotation [a]20/D +21.0 to +25.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR