(R)-(+)-2-(4-Hydroxyphenoxy)propionic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: (R)-2-(4-Hydroxyphenoxy)propanoic acid

H0937-5G
TCIAH0937-5GEA 485 NOK
TCIAH0937-5G
(R)-(+)-2-(4-Hydroxyphenoxy)propionic acid ≥98.0% (by GC, titration analysis)
(R)-(+)-2-(4-Hydroxyphenoxy)propionic acid
Formel: C₉H₁₀O₄
MW: 182.18 g/mol
MDL nummer: MFCD00274088
CAS-nummer: 94050-90-5
EINECS: 407-960-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ad9ad4c5ebb59cea3bf82b7d7930b967f619f48&vid=8ad9ad4c5ebb59cea3bf82b7d7930b967f619f48&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +36.0 to +39.0 deg (C=0.1, MeOH)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR