Methyl (R)-(+)-2-(4-hydroxyphenoxy)propionate ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: (R)-(+)-2-(4-Hydroxyphenoxy)propionic Acid Methyl Ester, (R)-Methyl 2-(4-hydroxyphenoxy)propanoate

H0956-25G
TCIAH0956-25GEA 2350 NOK
TCIAH0956-25G
Methyl (R)-(+)-2-(4-hydroxyphenoxy)propionate ≥98.0% (by GC)
Methyl (R)-(+)-2-(4-hydroxyphenoxy)propionate
Formel: C₁₀H₁₂O₄
MW: 196.20 g/mol
MDL nummer: MFCD00274087
CAS-nummer: 96562-58-2
EINECS: 411-950-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 67.0 deg-C
Specific rotation [a]20/D +40.0 to +45.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR