4-(Hydroxymethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-(Hydroxymethyl)cyclohexanecarboxylic acid, 4-Carboxy-1-cyclohexanemethanol

H1015-25G H1015-5G
TCIAH1015-25GEA 5800 NOK
TCIAH1015-25G TCIAH1015-5G
4-(Hydroxymethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
4-(Hydroxymethyl)cyclohexanecarboxylic acid (cis and trans mixture)
Formel: C₈H₁₄O₃
MDL nummer: MFCD01734677
CAS-nummer: 13380-84-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9858d2d968e6f0ea3361d29ded06aa102f8f431c&vid=9858d2d968e6f0ea3361d29ded06aa102f8f431c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Purple powder to crystal
Purity (GC) min. 98.0 % (total of isomers)
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR